Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Classroom Clock Kit
SCI_ED

Catalog No. S79398
Change view
Click to view available options
Grade:
Pre-K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79398 Pre-K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79398 Supplier Learning Resources, Inc Supplier No. LER 2102
Add to Cart
Add to Cart

Includes

  • Big Time™ 12-Hour Demonstration Clock
  • 4 in. geared mini-clocks (24 each)
  • Teacher's Guide

Additional mini-clocks available.

Order Info

  • Grades PK-4
  • Additional mini-clocks available

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
Format Hands-on Learning
Product Type Classroom Clock Kit
Includes Big Time™ 12 Hours demonstration clock, 4 in. geared mini clocks, teachers Guide
For Use With (Application) Demonstrate time-telling concepts while students follow along in hands-on activities
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.