Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Classroom DNA Discovery LabStation™
SDPSCI_ED

Catalog No. S72163 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
6 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S72163 6 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72163 Supplier Edvotek™ Supplier No. 5070
Add to Cart
Add to Cart

Newly reengineered M36 Electrophoresis Apparatus features the latest in electrophoresis design and supports up to six groups of students.

  • Includes the following:
  • 1 x Cat. #515 Edvotek™ M36 HexaGel Electrophoresis Apparatus (7 x 7 cm Trays (6))
  • 1 x Cat. #507 Edvotek™ DuoSource™ 75 Power Supply
  • 2 x Cat. #588 Fixed Volume MiniPipette™ 40 μL
  • 1 x Cat. #636-1 Yellow Micropipet Tips 1 to 200 μL (1 Rack of 96 Tips)
  • 1 x Cat. #S-51 Whose DNA Was Left Behind

Specifications

Grade 7 to Undergraduate
Age 14 to 20
For Use With (Application) DNA Analysis
Time Range Results in 30 to 40 minutes
Class Size 6 Lab Groups
Product Type LabStation™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.