Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Classroom DNA Electrophoresis LabStation™
SDPSCI_ED

Catalog No. S68645 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
24 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S68645 24 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68645 Supplier Edvotek™ Supplier No. 5062
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

(1) M36 HexaGel™ Electrophoresis Apparatus (Six 7 x 7 cm Trays) (1) DuoSource™ 150 (75/150 V for 1 or 2 units) (2) Fixed Volume MiniPipet (40 μl) (1) Yellow Micropipet Tips (1 - 200 μl / 2 Racks of 96) (1) DNA Fingerprinting Classroom Kit

The perfect way to bring DNA electrophoresis to your whole class! This LabStation™ provides all you need to run any DNA or dye electrophoresis kit with your students.

Includes:

  • M36 HexaGel™ Electrophoresis Apparatus (Six 7 x 7 cm Trays)
  • DuoSource™ 150 (75/150 V for 1 or 2 units)
  • 2 x Fixed Volume MiniPipette (40 μL)
  • Yellow Micropipet Tips (1-200 μL / 2 racks of 96)
  • DNA Fingerprinting Classroom Kit

Specifications

Grade 6 to 8
Includes M36 HexaGel™ Electrophoresis Apparatus, six 7 x 7 cm Trays (Mfr. No. 515 ; DuoSource™ 150 Power Supply (Mfr. No.509 ); 2 Fixed Volume MiniPipette, 40 μL (Mfr. 588); Yellow Micropipet Tips, 1 to 200 μL, 2 Racks of 96 (Mfr. No. 636 ); DNA Fingerprinting Classroom Kit (Mfr. No. 130 )
Class Size 24 Students
Product Type LabStation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.