Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Classroom PCR Discovery LabStation™
SDPSCI_ED

Catalog No. S72164 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
1 to 2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S72164 1 to 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72164 Supplier Edvotek™ Supplier No. 5072
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

A versatile package suited to run the full spectrum of horizontal electrophoresis experiments.

  • Includes the following:
  • 6 x Cat. #502/504 Edvotek™ M12 Complete™ Package (7 x 14 cm Tray & 7 x 7 cm Trays (2)
  • 3 x Cat. #509 Edvotek™ DuoSource™ 150 (75/150 V, for 1 or 2 units)
  • 6 x Cat. #590 Variable Micropipette (5 to 50 μL)
  • 2 x Cat. #534 Piccolo Microcentrifuge
  • 1 x Cat. #541-542 Edvotek™ EdvoCycler™ 2 (48 x 0.2 mL)
  • 1 x Cat. #557 Edvotek™ TruBlu™ Blue Light Transilliminator (14.5 x 18 cm viewing surface)
  • 1 x Cat. #539 Edvotek™ 1.8 L Water Bath

Specifications

Grade 8 to Undergraduate
Age 14 to 20
For Use With (Application) Biotechnology Education
Time Range Results in 30 to 40 minutes
Class Size 1 to 2 Lab Groups
Product Type LabStation™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.