Learn More
AdipoGen clasto-Lactacystin beta-lactone (100 µg)
SDP
Supplier:  AdipoGen AGCN20442C100
Encompass_Preferred
Chemical. CAS: 154226-60-5. Formula: C10H15NO4. MW: 213.2. Potent and selective irreversible and cell permeable proteasome inhibitor. Inhibits the chymotrypsin-like, trypsin-like and caspase-like peptidase activity of the proteasome. The active metabolite of lactacystin (Prod. No. AG-CN2-0104) with higher potency since it does not require hydrolysis in order to become cell permeable. Calpain and cathepsin inhibitor. Apoptosis inducer. Anticancer compound. Induces differentiation and inhibits cell cycle progression in several tumor cell lines. Induces neuritogenesis. Autophagy inducer.
Catalog No. 50-205-3609