Learn More
Medchemexpress LLC Clathrin-IN-4 | 762242-59-1 | C17H18Br2N2O | 1 ML
SDP
Catalog No. 5000373918
Encompass_Preferred
Clathrin-IN-4 is a potent inhibitor of clathrin-mediated endocytosis and also acts as a dynamin I GTPase activity inhibitor. It is an analogue of Wiskostatin and appears as a white to off-white solid.
  • Potent inhibitor of clathrin-mediated endocytosis with an IC50 of 2.1 μM
  • Inhibits dynamin I GTPase activity with an IC50 of 9.1 μM
  • Appears as a white to off-white solid
Catalog No. 50-003-73918 Supplier Medchemexpress LLC Supplier No. HY15221810MM/1ML
Add to Cart
Add to Cart
Clathrin-IN-4 is a potent inhibitor of clathrin-mediated endocytosis and also acts as a dynamin I GTPase activity inhibitor. It is an analogue of Wiskostatin and appears as a white to off-white solid.
  • Potent inhibitor of clathrin-mediated endocytosis with an IC50 of 2.1 μM
  • Inhibits dynamin I GTPase activity with an IC50 of 9.1 μM
  • Appears as a white to off-white solid