Learn More
Medchemexpress LLC Claudin 18.2 antibody | 10 UL
SDP
Catalog No. 5000441237
Encompass_Preferred
This rabbit-derived, non-conjugated IgG polyclonal antibody targets Claudin 18.2 for research use. It is affinity purified and supplied in 1*PBS (pH 7.4), 0.2% BSA, 50% Glycerol, with 0.05% Sodium Azide as a preservative. It should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Targets Claudin 18.2
  • For research use only
  • Affinity purified
  • Supplied in PBS, BSA, and Glycerol
  • Contains Sodium Azide as a preservative
  • Observe 35 kDa molecular weight
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles
Catalog No. 50-004-41237 Supplier Medchemexpress LLC Supplier No. HYP8177310UL
This rabbit-derived, non-conjugated IgG polyclonal antibody targets Claudin 18.2 for research use. It is affinity purified and supplied in 1*PBS (pH 7.4), 0.2% BSA, 50% Glycerol, with 0.05% Sodium Azide as a preservative. It should be stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles.
  • Targets Claudin 18.2
  • For research use only
  • Affinity purified
  • Supplied in PBS, BSA, and Glycerol
  • Contains Sodium Azide as a preservative
  • Observe 35 kDa molecular weight
  • Stable for 1 year when stored at -20°C
  • Avoid repeated freeze/thaw cycles