Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Claudin 18.2 Antibody | 50 UL
SDP

Catalog No. 5000441304
Encompass_Preferred

Claudin 18.2 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use only. It targets Claudin 18.2 and reacts with human, mouse, and rat species. This antibody is suitable for various applications including Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), Immunohistochemistry-Paraffin (IHC-P), and Flow Cytometry (FC).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 1*PBS (pH7.4), 0.2% BSA, 50% Glycerol. Preservative: 0.05% Sodium Azide.
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Applications: WB, ICC/IF, IHC-P, FC

Catalog No. 50-004-41304 Supplier Medchemexpress LLC Supplier No. HYP8177350UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Claudin 18.2 Antibody is a rabbit-derived, non-conjugated IgG polyclonal antibody designed for research use only. It targets Claudin 18.2 and reacts with human, mouse, and rat species. This antibody is suitable for various applications including Western Blot (WB), Immunocytochemistry/Immunofluorescence (ICC/IF), Immunohistochemistry-Paraffin (IHC-P), and Flow Cytometry (FC).

  • Host: Rabbit
  • Clonality: Polyclonal
  • Species reactivity: Human, mouse, rat
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG
  • Appearance: Liquid
  • Formulation: Supplied in 1*PBS (pH7.4), 0.2% BSA, 50% Glycerol. Preservative: 0.05% Sodium Azide.
  • Storage and stability: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles.
  • Applications: WB, ICC/IF, IHC-P, FC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.