Learn More
OriGene Claudin 4 (CLDN4) CytoSection
SDP
Catalog No. NC3881577
Encompass_Preferred
Transient overexpression of CLDN4 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC3881577 Supplier OriGene Supplier No. TS400490P5
Transient overexpression of CLDN4 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack