Learn More
Biomatik Corporation Claudin-18.2 Recombinant Antibody
Supplier:  Biomatik Corporation RAC07073
Encompass_Preferred
Claudin-18.2 Recombinant Antibody is a Human (Homo sapiens) Recombinant Antibody to CLDN18. The Claudin-18.2 Recombinant Antibody has been validated for the following applications: ELISA, FC. Host: HEK293F Cell. Species: Human (Homo sapiens). Immunogen: Recombinant Human Claudin-18.2 Protein. Applications: ELISA, FC. Target Synonyms: CLDN18. Accession Number: P56856-2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6389