Learn More
ABclonal Technology Claudin18.2 Rabbit mAb
SDP
Catalog No. 5000013689
Encompass_Preferred
This gene encodes a member of the claudin family Claudins are integral membrane proteins and components of tight junction strands Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets and also play critical roles in maintaining cell polarity and signal transductions This gene is upregulated in patients with ulcerative colitis and highly overexpressed in infiltrating ductal adenocarcinomas PKC MAPK AP-1 (protein kinase C mitogen-activated protein kinase activator protein-1) dependent pathway regulates the expression of this gene in gastric cells Alternatively spliced transcript variants encoding different isoforms have been identified
Catalog No. 50-000-13689 Supplier ABclonal Technology Supplier No. A2782220UL
This gene encodes a member of the claudin family Claudins are integral membrane proteins and components of tight junction strands Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets and also play critical roles in maintaining cell polarity and signal transductions This gene is upregulated in patients with ulcerative colitis and highly overexpressed in infiltrating ductal adenocarcinomas PKC MAPK AP-1 (protein kinase C mitogen-activated protein kinase activator protein-1) dependent pathway regulates the expression of this gene in gastric cells Alternatively spliced transcript variants encoding different isoforms have been identified