Learn More
Sigma Aldrich Fine Chemicals Biosciences ClaviesR biohazard autocla
Catalog No. 502900793
Encompass
Soft pliant all-cotton terry cloth gloves are excellent for handling hot objects out of an autoclave or oven. Gauntlets available in two lengths and provide wrist and forearm protection.Heat-resistant up to 232C (450f)Can replace hazardous asbestos glovesProvides wrist and forearm protectionBright orange color offers high visibilityMachine wash air dry
Catalog No. 50-290-0793 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z4084921PAK
Soft pliant all-cotton terry cloth gloves are excellent for handling hot objects out of an autoclave or oven. Gauntlets available in two lengths and provide wrist and forearm protection.Heat-resistant up to 232C (450f)Can replace hazardous asbestos glovesProvides wrist and forearm protectionBright orange color offers high visibilityMachine wash air dry