Learn More
Medchemexpress LLC Clavulanate lithium | 61177-44-4 | 99.9% | 205.09 | C8H8LiNO5 | 10 MG
SDP
Catalog No. 5000298193
Encompass_Preferred
Clavulanate lithium is a β-lactamase inhibitor supplied as a high-purity analytical standard for laboratory research. It exhibits weak antibacterial activity alone but prevents inactivation of β-lactam antibiotics by microbial β-lactamases; intended for analytical and mechanistic studies. Not for clinical or human use.
  • High purity (99.9%) appropriate for analytical applications.
  • Chemical formula C8H8LiNO5; molecular weight 205.09 g/mol.
  • Available in small, defined pack sizes for precise dosing and assays.
  • Provided with manufacturer documentation and batch-specific certificates of analysis.
  • Intended for research use only; not for therapeutic use.
Catalog No. 50-002-98193 Supplier Medchemexpress LLC Supplier No. HYA0256B10MG
Clavulanate lithium is a β-lactamase inhibitor supplied as a high-purity analytical standard for laboratory research. It exhibits weak antibacterial activity alone but prevents inactivation of β-lactam antibiotics by microbial β-lactamases; intended for analytical and mechanistic studies. Not for clinical or human use.
  • High purity (99.9%) appropriate for analytical applications.
  • Chemical formula C8H8LiNO5; molecular weight 205.09 g/mol.
  • Available in small, defined pack sizes for precise dosing and assays.
  • Provided with manufacturer documentation and batch-specific certificates of analysis.
  • Intended for research use only; not for therapeutic use.