Learn More
Medchemexpress LLC Clavulanate lithium | 61177-44-4 | 99.9% | C8H8LiNO5 | 5 MG
SDP
Catalog No. 5000432467
Encompass_Preferred
Clavulanate lithium is a potent beta-lactamase inhibitor that also acts as an antibiotic. While exhibiting weak antibacterial activity alone, it prevents the inactivation of beta-lactam antibiotics by microbial lactamase when combined with them. It has a molecular weight of 205.09 and an appearance of a solid, white to off-white color.
  • Potent beta-lactamase inhibitor
  • Acts as an antibiotic
  • Prevents inactivation of beta-lactam antibiotics
  • Solid, white to off-white appearance
  • Soluble in H2O and DMSO
Catalog No. 50-004-32467 Supplier Medchemexpress LLC Supplier No. HYA0256B5MG
Clavulanate lithium is a potent beta-lactamase inhibitor that also acts as an antibiotic. While exhibiting weak antibacterial activity alone, it prevents the inactivation of beta-lactam antibiotics by microbial lactamase when combined with them. It has a molecular weight of 205.09 and an appearance of a solid, white to off-white color.
  • Potent beta-lactamase inhibitor
  • Acts as an antibiotic
  • Prevents inactivation of beta-lactam antibiotics
  • Solid, white to off-white appearance
  • Soluble in H2O and DMSO