Learn More
Medchemexpress LLC Clavulanate potassium | 61177-45-5 | MFCD01710901 | 99.5% | 237.25 | C8H8KNO5 | 10 MG
SDP
Catalog No. 5000298778
Encompass_Preferred
Clavulanate potassium is the potassium salt of clavulanic acid, a potent β-lactamase inhibitor used in research to study antibiotic resistance and to enhance the activity of β-lactam antibiotics. It is supplied as a high-purity solid for in vitro assays, analytical testing, and formulation research.
  • Potent β-lactamase inhibitor for resistance studies.
  • Used to potentiate β-lactam antibiotics in combination assays.
  • High purity suitable for analytical and in vitro applications.
  • Well-defined chemical identity (CAS 61177-45-5; MW 237.25).
  • Available in small research pack sizes for laboratory use.
Catalog No. 50-002-98778 Supplier Medchemexpress LLC Supplier No. HYA0256A10MG
Clavulanate potassium is the potassium salt of clavulanic acid, a potent β-lactamase inhibitor used in research to study antibiotic resistance and to enhance the activity of β-lactam antibiotics. It is supplied as a high-purity solid for in vitro assays, analytical testing, and formulation research.
  • Potent β-lactamase inhibitor for resistance studies.
  • Used to potentiate β-lactam antibiotics in combination assays.
  • High purity suitable for analytical and in vitro applications.
  • Well-defined chemical identity (CAS 61177-45-5; MW 237.25).
  • Available in small research pack sizes for laboratory use.