Learn More
Medchemexpress LLC Clavulanic acid | 58001-44-8 | 99.45% | C8H9NO5 | 5 MG
SDP
Catalog No. 5000432267
Encompass_Preferred
Clavulanic acid is a naturally occurring, powerful bacterial β-lactamase inhibitor used for research into infections caused by bacteria, including ear infections.
  • Naturally occurring β-lactamase inhibitor
  • Effective against a wide spectrum of gram-positive and gram-negative bacteria
Catalog No. 50-004-32267 Supplier Medchemexpress LLC Supplier No. HYA02565MG
Add to Cart
Add to Cart
Clavulanic acid is a naturally occurring, powerful bacterial β-lactamase inhibitor used for research into infections caused by bacteria, including ear infections.
  • Naturally occurring β-lactamase inhibitor
  • Effective against a wide spectrum of gram-positive and gram-negative bacteria