Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clavulanic Acid Potassium with Cellulose (1:1), G-Biosciences
SCI_ED

Catalog No. S973249
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S973249 5 g
S973245 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S973249 Supplier G-Biosciences Supplier No. RC977
Add to Cart
Add to Cart

Clavulanic acid potassium (clavulanate potassium) is a Beta-lactamase inhibitor isolated from Streptomyces clavuligerus, which irreversibly inactivates Beta-lactamase enzymes of Beta-lactam resistant organisms. This prevents these organisms from degrading Beta-lactam antibiotics.

Clavulanic acid can be combined with antibiotics to increase their potency and is often combined with amoxicillin.

Specifications

Chemical Name or Material Clavulanic Acid Potassium
CAS 61177-45-5, 9004-34-6
Physical Form Flakes or Powder
Quantity 5 g
Molecular Weight (g/mol) 237.25 g/mol
Percent Purity 48.0-53.5%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.