Learn More
Cayman Chemical Clavulanate (potassium salt), Purity: ≥95%, CAS: 61177-45-5, 50 mg
SDP
Catalog No. 502983055
Encompass_Preferred
A β-lactamase inhibitor that is effective against Ambler class A β-lactamases (IC50 values range from 12 to 60 nM); commonly used with other antibiotics that would be inactivated by β-lactamases
Catalog No. 50-298-3055 Supplier Cayman Chemical Supplier No. 1945650 MG
A β-lactamase inhibitor that is effective against Ambler class A β-lactamases (IC50 values range from 12 to 60 nM); commonly used with other antibiotics that would be inactivated by β-lactamases