Learn More
Bioworld CLC/Ipomoea Basal Medium CP w/ Vitamins, 50 L
Catalog No. 50254572
Encompass_Preferred
CLC/Ipomoea Basal Medium CPFor embryogenic Callus GrowthContains Macronutrients, Micronutrients, and Vitamins Provided as 1 unit to make 10L (30630011-1), 25L or 50L of medium, or as 10 units (30630011-2) that each make 1L of mediumWorking Concentration: 6.7 g/L Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-572 Supplier Bioworld Supplier No. 306300114
CLC/Ipomoea Basal Medium CPFor embryogenic Callus GrowthContains Macronutrients, Micronutrients, and Vitamins Provided as 1 unit to make 10L (30630011-1), 25L or 50L of medium, or as 10 units (30630011-2) that each make 1L of mediumWorking Concentration: 6.7 g/L Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested