Learn More
Bioworld CLC/Ipomoea Basal Medium EP w/ Vitamins, 25 L
Catalog No. 50254575
Encompass_Preferred
CLC/Ipomoea Basal Medium EPFor embryogenic developmentContains Macronutrients, Micronutrients, and Vitamins Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L (30630012-1), 25L or 50L of medium, or as 10 units (30630012-2) that each make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-575 Supplier Bioworld Supplier No. 306300123
CLC/Ipomoea Basal Medium EPFor embryogenic developmentContains Macronutrients, Micronutrients, and Vitamins Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L (30630012-1), 25L or 50L of medium, or as 10 units (30630012-2) that each make 1L of mediumPlant tissue culture tested