Learn More
Aviva Systems Biology Corp CLCNKA Antibody - N-terminal region (ARP35132_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35132_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLCNKA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLCNKA. Alternate names: CLCK1, ClC-K1, MGC61490, hClC-KaApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat
Catalog No. 50-014-55650