Learn More
Aviva Systems Biology Corp CLCNKB Antibody - C-terminal region (ARP34917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLCNKB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLCNKB. Alternate names: CLCKB, ClC-K2, ClC-Kb, hClC-KbApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55607