Learn More
Aviva Systems Biology Corp CLDN1 Antibody - C-terminal region (ARP33623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN1. Alternate names: CLD1, SEMP1, ILVASCApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-55098