Learn More
Aviva Systems Biology Corp CLDN10 Antibody - C-terminal region (ARP33614_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33614_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN10. Alternate names: OSP-L, CPETRL3ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55095