Learn More
Aviva Systems Biology Corp CLDN16 Antibody - C-terminal region (ARP33632_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33632_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN16. Alternate names: HOMG3, PCLN1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55102