Learn More
Aviva Systems Biology Corp CLDN17 Antibody - middle region (ARP33615_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33615_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN17. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human CLDN17. Alternate names: ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-52912