Learn More
Aviva Systems Biology Corp CLDN18 Antibody - middle region (ARP33612_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33612_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN18. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLDN18. Alternate names: SFTA5, SFTPJApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52911