Learn More
Biomatik Corporation Cldn18 Polyclonal Antibody
Supplier:  Biomatik Corporation CAC10906
Encompass_Preferred
Cldn18 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to CLDN18. The Cldn18 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Claudin-18 Protein (20-80Aa). Applications: ELISA, WB, IHC. Target Synonyms: CLDN18. Accession Number: P56856. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7133