Learn More
Aviva Systems Biology Corp CLDN19 Antibody - C-terminal region (ARP33619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN19. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN19. Alternate names: HOMG5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55096