Learn More
Aviva Systems Biology Corp CLDN2 Antibody - N-terminal region (ARP42845_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42845_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLDN2. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-58349