Learn More
ABclonal Technology CLDN2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A140851000UL
Encompass_Preferred
This gene product belongs to the claudin protein family whose members have been identified as major integral membrane proteins localized exclusively at tight junctions. Claudins are expressed in an organ-specific manner and regulate tissue-specific physiologic properties of tight junctions. This protein is expressed in the intestine. Alternatively spliced transcript variants with different 5 untranslated region have been found for this gene.
Catalog No. 50-001-09042