Learn More
Aviva Systems Biology Corp CLDN23 Antibody - C-terminal region (ARP33633_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33633_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN23. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN23. Alternate names: CLDNL, hCG1646163ApplicationIHC, WB. Species reactivity: Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-52915