Learn More
Aviva Systems Biology Corp Cldn3 Antibody - C-terminal region (ARP42831_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42831_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cldn3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI182374, Cpetr2, mRVP1ApplicationWB. Species reactivity: Cow, Dog, Mouse, Rat
Catalog No. 50-285-032