Learn More
Aviva Systems Biology Corp CLDN5 Antibody - C-terminal region (ARP33627_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33627_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CLDN5. Alternate names: AWAL, BEC1, CPETRL1, TMVCFApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55099