Learn More
Aviva Systems Biology Corp CLDN7 Antibody - middle region (ARP33613_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33613_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDN7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLDN7. Alternate names: CEPTRL2, CPETRL2, Hs.84359, claudin-1, CLDN-7ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-55094