Learn More
Aviva Systems Biology Corp CLDND1 Antibody - middle region (ARP44926_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44926_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLDND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CLDND1. Alternate names: C3orf4, GENX-3745, MGC111162, MGC3316, MGC9861ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53439