Learn More
Air Science Usa Clean Bench 36" Wide
SDP
Catalog No. NC3893977
Encompass_Preferred
Purair LF Series Horizontal Laminar Flow bench, UV Option,36" / 900mm Nominal Width, Extra Tall Internal Height, 115V 60Hz, North American Cord set (unless specified).
Catalog No. NC3893977 Supplier Air Science Usa Supplier No. HLF36XTAUV
Purair LF Series Horizontal Laminar Flow bench, UV Option,36" / 900mm Nominal Width, Extra Tall Internal Height, 115V 60Hz, North American Cord set (unless specified).