Learn More
Clean Clay Loam, MilliporeSigma™ Supelco™
Catalog No. 111001668 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
Catalog No. Quantity
11-100-1668 100 g
1 options
Catalog No. 11-100-1668 Supplier MilliporeSigma Supelco Supplier No. CLNSOIL2100G
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Clean clay loam may be used as a reference material in the recovery of nitro-organic explosives in soil samples using gas chromatography coupled with electron capture detector (GC-ECD).
Featured Industries: Environmental
TRUSTED_SUSTAINABILITY
Specifications
CAS 14464-46-1
Type Certified Reference Material
Quantity 100 g