Learn More
Clean Collect™ Blood Culture Kits
Catalog No. 23200104 Shop All Siemens Healthineers Products
Change view
Click to view available options
Includes K-Shield butterfly needle; Compatible with BacT/Alert Systems
Catalog No.
23-200-104 Includes K-Shield butterfly needle; Compatible with BacT/Alert Systems
1 options
Catalog No. 23-200-104 Supplier Siemens Healthcare Diagnostics Supplier No. 11416778
Add to Cart
Add to Cart
Blood culture diversion system prevents contamination in peripheral blood cultures
Reducing blood culture contamination rates prevents unnecessary treatment of patients. Clean Collect kits can help to increase quality and save considerable resources.
  • Prevents blood culture contamination using proprietary diversion method
  • Average contamination rate of 0.5% compared to 3% national average
  • Diversion method reduces contamination rates by an average of 74%
  • Standardized kits promote consistency for long term improvement
  • Reduces hospital costs considerably
  • Ambient temperature (0-40°C) dry storage; 10 month shelf life