Learn More
Clean Collect™ Diversion IV Blood Culture Collection Kit
SDP
Catalog No. 23200123
Change view
Click to view available options
Quantity:
40/Cs.
Catalog No. Quantity
23-200-123 40/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 23-200-123 Supplier LSL Healthcare Inc Supplier No. CC315SM
Add to Cart
Add to Cart
This item is not returnable. View return policy
Prevent blood culture contamination with a diversion system peripheral blood culture kit.
  • Reduces contamination
  • Improves patient outcomes
  • Saves hospital resources
  • Includes the following: sterile diversion tube (red top), chloraprep antiseptic, IV dressing, gauze, sterile gloves, tourniquet, alcohol pads, tracking decals, and instruction card
Specifications
Content And Storage 5°C to 25°C
Quantity 40/Cs.