Learn More
Research Products International Corp Clean Low Retention Tips 10ul
SDP
Catalog No. 502946125
Encompass
10µl, Graduated at 2 & 10 µl, Fit P2, P10 Pipetman RPI Clean Low Retention Tips feature a specially formulated NoStick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision. EcoPac Refill System also provides convenient lift-and-load refilling and reduces lab waste by 70%. EcoPac refill trays are robust withstand multiple autoclave cycles.
Catalog No. 50-294-6125 Supplier Research Products International Corp Supplier No. 147204
10µl, Graduated at 2 & 10 µl, Fit P2, P10 Pipetman RPI Clean Low Retention Tips feature a specially formulated NoStick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision. EcoPac Refill System also provides convenient lift-and-load refilling and reduces lab waste by 70%. EcoPac refill trays are robust withstand multiple autoclave cycles.