Learn More
Research Products International Corp Clean Low Retention Tips 10ul
SDP
Supplier:  Research Products International Corp 147201
Encompass
10µl, Graduated at 2 & 10 µl, Fit P2, P10 Pipetman RPI Clean Low Retention Tips feature a specially formulated NoStick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision. Racks are uniquely designed to facilitate one hand operation.
Catalog No. 50-294-6115