Learn More
Research Products International Corp Clean Low Retention Tips 200ul
SDP
Catalog No. 502946127
Encompass
200 µl, Graduated at 10, 50, 100 µl RPI Clean Low Retention Tips feature a specially formulated NoStick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision.
Catalog No. 50-294-6127 Supplier Research Products International Corp Supplier No. 147210
200 µl, Graduated at 10, 50, 100 µl RPI Clean Low Retention Tips feature a specially formulated NoStick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision.