Learn More
Research Products International Corp Clean Low Retentn Tips 1000ul
SDP
Supplier:  Research Products International Corp 147216
Encompass
1000 µl, Graduated at 100, 500 and 1000 µl RPI Clean Low Retention Tips feature a specially formulated No Stick resin that offers a highly hydrophobic inner surface. Unique RPI Clean Tips offer a much cleaner release, provide less fluid retention, less sample loss, and eliminate the need to touch off. Universal fit tips allow complete fluid control when working with smaller volumes and promote complete pipetter accuracy and precision.
Catalog No. 50-294-6122