Learn More
Uline Clean Mat Sheets with Frame - 24 by 36 - ULINE - Quantity: 1
Catalog No. NC1459802
Encompass
Lint-free 24 x 36 inch blue polyester cleanroom mat sheet with pressure sensitive adhesive backing designed for disposable replacement to maintain cleanroom floor cleanliness by collecting dirt and debris from foot traffic when placed in plastic frame adhered to floor.
Catalog No. NC1459802 Supplier Uline Supplier No. H1567BLU
Lint-free 24 x 36 inch blue polyester cleanroom mat sheet with pressure sensitive adhesive backing designed for disposable replacement to maintain cleanroom floor cleanliness by collecting dirt and debris from foot traffic when placed in plastic frame adhered to floor.