Learn More
MSC Chapin Sprayer, 1 Gal, Chemical Safe, Polyethylene Tank, 1 ea
Catalog No. 502867191
Encompass
The Chapin 25012 is a 1-gal. handheld sprayer designed for deck and yard applications. It features a translucent polyethylene tank with a funnel top, facilitating easy filling and cleaning. This sprayer is compatible with a range of cleaners, sealers, and transparent stains.
  • Capacity of 1 gal.
  • Polyethylene tank material
  • Funnel top for easy filling and cleaning
  • Reinforced hose
  • Chemical safe for various cleaners and stains
  • Ergonomic handle for comfortable pumping and carrying
  • Total control spray shield to protect siding
  • Includes three poly fan nozzles for fine, medium, and coarse spray patterns
  • In-tank filter for consistent operation
  • Wand length of 12 inch
Catalog No. 50-286-7191 Supplier MSC Supplier No. 02399392
The Chapin 25012 is a 1-gal. handheld sprayer designed for deck and yard applications. It features a translucent polyethylene tank with a funnel top, facilitating easy filling and cleaning. This sprayer is compatible with a range of cleaners, sealers, and transparent stains.
  • Capacity of 1 gal.
  • Polyethylene tank material
  • Funnel top for easy filling and cleaning
  • Reinforced hose
  • Chemical safe for various cleaners and stains
  • Ergonomic handle for comfortable pumping and carrying
  • Total control spray shield to protect siding
  • Includes three poly fan nozzles for fine, medium, and coarse spray patterns
  • In-tank filter for consistent operation
  • Wand length of 12 inch