Learn More
Yamato Scientific America, Inc. Yamato 327L Clean Oven, Large Capacity, Maximum Temp at 360°C, 220V 3 phase
Catalog No. 501898042 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Space-saving large capacity 327L forced convection Class 100 clean oven. Improved visibility and operability of control panel. Stable cleanliness through forced circulation with rear exhaust. Displays power consumption, CO2 emissions and heater manipulated variables on the control panel. Incorporates a maximum of 99 steps, 99 patterns program controller with repeat function. Offers several options such as recorder, manual/auto damper, N2 gas introducer with flow meter and emergency switch. Operating temp range of ambient +30~360°C
Catalog No. 50-189-8042 Supplier Yamato Scientific America, Inc. Supplier No. DTS830
Add to Cart
Add to Cart
Request a Quote
Space-saving large capacity 327L forced convection Class 100 clean oven. Improved visibility and operability of control panel. Stable cleanliness through forced circulation with rear exhaust. Displays power consumption, CO2 emissions and heater manipulated variables on the control panel. Incorporates a maximum of 99 steps, 99 patterns program controller with repeat function. Offers several options such as recorder, manual/auto damper, N2 gas introducer with flow meter and emergency switch. Operating temp range of ambient +30~360°C