Learn More
Yamato Scientific America, Inc. Yamato 91L Clean Oven, Maximum Temp at 360°C, 220V 3 phase
Catalog No. 501898039 Shop All Yamato Scientific America Inc. Products
Encompass_Preferred
Forced convection 91L Class 100 clean oven suitable for temperature test in a dust-free environment. Improved visibility and operability with its V type controller. Displays power consumption, CO2 emissions and heater manipulated variables on the control panel. Adopts anti-fouling casters which prevents wheel contamination during transportation. Improved visibility of HEPA filter replacement timing by three color indication. Enhanced safety with its phase-reversal relay detecting incorrect power source at installation Larger cable port of φ33mm. Improved optional accessories and more customization options. Operating temp range of ambient +30~360°C
Catalog No. 50-189-8039 Supplier Yamato Scientific America, Inc. Supplier No. DT411
Add to Cart
Add to Cart
Request a Quote
Forced convection 91L Class 100 clean oven suitable for temperature test in a dust-free environment. Improved visibility and operability with its V type controller. Displays power consumption, CO2 emissions and heater manipulated variables on the control panel. Adopts anti-fouling casters which prevents wheel contamination during transportation. Improved visibility of HEPA filter replacement timing by three color indication. Enhanced safety with its phase-reversal relay detecting incorrect power source at installation Larger cable port of φ33mm. Improved optional accessories and more customization options. Operating temp range of ambient +30~360°C