Learn More
TrueCare Biomedix Sterile Clean Room Gown, 40gsm+-3gsm White SMS,anti-static treated, stand up collar with velcro closure in the back, with elastic wrists, 1pcsterile bag,50pcsctn, R
Catalog No. NC2217185
Encompass_Preferred
Sterile Clean Room Gown, 40gsm+-3gsm White SMS,anti-static treated, stand up collar with velcro closure in the back, with elastic wrists, 1pcsterile bag,50pcsctn, R
This item is not returnable. View return policy
Catalog No. NC2217185 Supplier TrueCare Biomedix Supplier No. TCBA40STR
Sterile Clean Room Gown, 40gsm+-3gsm White SMS,anti-static treated, stand up collar with velcro closure in the back, with elastic wrists, 1pcsterile bag,50pcsctn, R