Learn More
TrueCare Biomedix Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals1pcsterile bag,50pcsctn, R
Catalog No. NC2217194
Encompass_Preferred
Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals1pcsterile bag,50pcsctn, R
This item is not returnable. View return policy
Catalog No. NC2217194 Supplier TrueCare Biomedix Supplier No. TCBA54STR
Sterile Clean Room Gown, 52gsm+-3gsm Breathable PEPP, Level 3 impervious, CHEMO TESTED stand up collar with velcro closure in the back, with elastic wrists, seam seals1pcsterile bag,50pcsctn, R